Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsy-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269216.3 | NP_001256145.1 | 13 - 14 | 3358 | 101 | |
NM_001269217.3 | NP_001256146.1 | 11 - 12 | 2496 | 101 | ||
NM_001269218.3 | NP_001256147.1 | 14 - 15 | 3483 | 101 | ||
NM_001276918.3 | NP_001263847.1 | 13 - 14 | 3348 | 101 | ||
NM_001276919.3 | NP_001263848.1 | 11 - 12 | 2496 | 101 | ||
NM_001276920.4 | NP_001263849.1 | 14 - 15 | 3483 | 101 | ||
NM_001373080.3 | NP_001359747.1 | 12 | 2717 | 101 | ||
NM_001373081.2 | NP_001360180.1 | 12 | 2698 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |