Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028662.4 | NP_001023833.1 | 3 - 4 | 314 | 83 | |
NM_001028663.6 | NP_001023834.1 | 6 - 7 | 1092 | 83 | ||
NM_001047640.4 | NP_001041105.1 | 4 - 5 | 856 | 83 | ||
NM_171533.6 | NP_741621.1 | 6 - 7 | 1092 | 83 | ||
GenBank mRNA | AF178536.1 | - | 6 - 7 | 1094 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |