Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047624.2 | NP_001041089.1 | 4 - 5 | 367 | 85 | |
NM_001047625.1 | NP_001041090.1 | 1 - 2 | 52 | 85 | ||
GenBank mRNA | AF535153.1 | - | 4 - 5 | 367 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |