Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136431.4 | NP_001129903.1 | 13 - 14 | 11534 | 65 | |
NM_001269453.3 | NP_001256382.1 | 11 - 12 | 11285 | 65 | ||
NM_171544.7 | NP_741632.2 | 14 - 15 | 11958 | 65 | ||
GenBank mRNA | AF053496.1 | - | 13 - 14 | 11534 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |