Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136431.4 | NP_001129903.1 | 3 - 4 | 487 | 77 | |
NM_001269453.3 | NP_001256382.1 | 1 - 2 | 238 | 77 | ||
NM_001269454.2 | NP_001256383.1 | 3 - 4 | 729 | 77 | ||
NM_001373124.2 | NP_001360523.1 | 2 | 796 | 77 | ||
NM_001383415.1 | 3 - 4 | 735 | 77 | |||
NM_171544.7 | NP_741632.2 | 4 - 5 | 911 | 77 | ||
GenBank mRNA | AF053496.1 | - | 3 - 4 | 487 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |