Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136431.4 | NP_001129903.1 | 8 - 9 | 7177 | 97 | |
NM_001269453.3 | NP_001256382.1 | 6 - 7 | 6928 | 97 | ||
NM_001269454.2 | NP_001256383.1 | 8 - 9 | 7419 | 97 | ||
NM_001373124.2 | NP_001360523.1 | 9 | 7486 | 97 | ||
NM_001383415.1 | 8 - 9 | 7425 | 97 | |||
NM_171544.7 | NP_741632.2 | 9 - 10 | 7601 | 97 | ||
GenBank mRNA | AF053496.1 | - | 8 - 9 | 7177 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |