Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269533.2 | NP_001256462.1 | 6 - 7 | 888 | 93 | |
NM_001269534.3 | NP_001256463.1 | 5 - 6 | 816 | 93 | ||
NM_001269535.3 | NP_001256464.1 | 4 - 5 | 666 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |