Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029058.4 | NP_001024229.1 | 7 - 8 | 1307 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |