Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029058.4 | NP_001024229.1 | 8 - 9 | 1441 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |