Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pro-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171594.5 | NP_741684.2 | 2 - 3 | 408 | 142 | |
GenBank mRNA | EU068466.1 | - | 2 - 3 | 400 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |