Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171612.7 | NP_741703.1 | 9 - 10 | 1019 | 71 | |
NM_171939.5 | NP_741704.1 | 9 - 10 | 1019 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |