Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270046.2 | NP_001256975.1 | 1 - 2 | 253 | 61 | |
NM_001374936.2 | NP_001362085.1 | 14 - 15 | 2352 | 61 | ||
NM_001374938.1 | NP_001362002.1 | 15 - 16 | 2425 | 61 | ||
NM_001380893.1 | NP_001367765.1 | 11 - 12 | 2133 | 61 | ||
NM_001383522.1 | 1 - 2 | 240 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |