Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncam-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171615.7 | NP_741707.3 | 10 - 11 | 2821 | 70 | |
NM_171616.5 | NP_741708.1 | 10 - 11 | 1705 | 70 | ||
NM_171617.5 | NP_741709.1 | 9 - 10 | 1845 | 70 | ||
GenBank mRNA | AF456360.2 | - | 10 - 11 | 1729 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |