Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029702.5 | NP_001024873.1 | 7 - 8 | 857 | 150 | |
NM_001374979.2 | NP_001362027.1 | 7 - 8 | 832 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |