Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373256.3 | NP_001359863.1 | 18 | 4236 | 84 | |
NM_001373257.3 | NP_001359864.1 | 17 - 18 | 4191 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon |