Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F35A5.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277025.4 | NP_001263954.1 | 3 - 4 | 357 | 66 | |
NM_171657.7 | NP_741756.1 | 4 - 5 | 483 | 66 | ||
NM_171658.5 | NP_741757.1 | 3 - 4 | 357 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |