Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F35A5.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171657.7 | NP_741756.1 | 7 - 8 | 753 | 135 | |
NM_171658.5 | NP_741757.1 | 6 - 7 | 627 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |