Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171659.6 | NP_741758.1 | 6 - 7 | 1232 | 143 | |
GenBank mRNA | U16367.1 | - | 6 - 7 | 1232 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |