Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373289.3 | NP_001359912.1 | 17 - 18 | 1918 | 93 | |
NM_001373290.4 | NP_001360675.1 | 14 - 15 | 1629 | 93 | ||
NM_171949.8 | NP_741779.1 | 15 - 16 | 1792 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |