Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373289.3 | NP_001359912.1 | 4 - 5 | 388 | 74 | |
NM_171949.8 | NP_741779.1 | 2 - 3 | 262 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |