Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373300.2 | NP_001360725.1 | 7 | 1164 | 69 | |
NM_001373301.2 | NP_001360721.1 | 7 | 1164 | 69 | ||
NM_001373304.2 | NP_001360724.1 | 8 | 1317 | 69 | ||
NM_001373305.2 | NP_001360720.1 | 8 | 1317 | 69 | ||
NM_171690.6 | NP_741796.2 | 8 - 9 | 1372 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |