Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnrr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373308.3 | NP_001360691.1 | 2 - 3 | 195 | 69 | |
NM_001373309.4 | NP_001359929.1 | 2 - 3 | 196 | 69 | ||
NM_171692.7 | NP_741798.1 | 2 - 3 | 194 | 69 | ||
NM_171954.7 | NP_741799.1 | 2 - 3 | 184 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |