Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pak-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029206.3 | NP_001024377.1 | 3 - 4 | 274 | 76 | |
NM_001029207.4 | NP_001024378.1 | 3 - 4 | 272 | 76 | ||
NM_001029208.5 | NP_001024379.1 | 2 - 3 | 126 | 76 | ||
GenBank mRNA | D83215.1 | - | 3 - 4 | 269 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |