Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029732.3 | NP_001024903.1 | 8 - 9 | 1288 | 85 | |
NM_001375044.2 | NP_001362132.1 | 7 - 8 | 1225 | 85 | ||
NM_171699.6 | NP_741807.2 | 9 - 10 | 1394 | 85 | ||
GenBank mRNA | AF407572.1 | - | 8 - 9 | 1288 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |