Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171712.5 | NP_741822.2 | 4 - 5 | 707 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |