Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhx-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029391.4 | NP_001024562.1 | 12 - 13 | 2199 | 118 | |
NM_001381015.1 | NP_001367515.1 | 12 - 13 | 2203 | 118 | ||
GenBank mRNA | AF497826.1 | - | 12 - 13 | 2221 | 118 | |
AF497827.1 | - | 9 - 10 | 1941 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |