Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171727.4 | NP_741838.1 | 5 - 6 | 831 | 83 | |
GenBank mRNA | AF082011.1 | - | 5 - 6 | 831 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |