Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cca-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381028.1 | NP_001367692.1 | 14 - 15 | 1637 | 94 | |
NM_001381029.1 | NP_001368577.1 | 14 - 15 | 1561 | 94 | ||
NM_001381030.1 | NP_001367691.1 | 14 - 15 | 1637 | 94 | ||
NM_001381031.2 | NP_001367690.1 | 14 - 15 | 1637 | 94 | ||
NM_171733.5 | NP_741848.1 | 16 - 17 | 1723 | 94 | ||
GenBank mRNA | AF368920.1 | - | 14 - 15 | 1586 | 94 | |
AY313898.1 | - | 14 - 15 | 1637 | 94 | ||
AY313899.1 | - | 14 - 15 | 1637 | 94 | ||
AY313900.1 | - | 14 - 15 | 1637 | 94 | ||
AY322480.1 | - | 14 - 15 | 1637 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |