Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnt-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373379.1 | NP_001360658.1 | 3 - 4 | 238 | 72 | |
NM_001373380.2 | NP_001359846.1 | 3 - 4 | 190 | 72 | ||
NM_001373381.2 | NP_001359848.1 | 2 - 3 | 2865 | 72 | ||
NM_001373382.2 | NP_001360657.1 | 2 - 3 | 2864 | 72 | ||
NM_001373383.1 | NP_001359847.1 | 2 - 3 | 2865 | 72 | ||
NM_001383610.1 | 2 - 3 | 109 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |