Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270129.4 | NP_001257058.1 | 1 - 2 | 143 | 67 | |
NM_001373393.3 | NP_001359981.1 | 1 - 2 | 141 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |