Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171743.6 | NP_741861.2 | 5 - 6 | 1256 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |