Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029399.4 | NP_001024570.1 | 10 - 11 | 1366 | 99 | |
NM_001029400.3 | NP_001024571.1 | 4 - 5 | 838 | 99 | ||
NM_001029401.5 | NP_001024572.1 | 4 - 5 | 602 | 99 | ||
GenBank mRNA | U29184.1 | - | 4 - 5 | 607 | 99 | |
U31537.1 | - | 10 - 11 | 1320 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |