Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171769.7 | NP_741893.1 | 3 - 4 | 416 | 62 | |
NM_171972.7 | NP_741894.1 | 3 - 4 | 419 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |