Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375119.3 | NP_001361905.1 | 22 | 5595 | 117 | |
NM_001375120.2 | NP_001361887.1 | 21 - 22 | 6141 | 117 | ||
NM_001375121.3 | NP_001361888.1 | 22 - 23 | 6042 | 117 | ||
NM_001375122.2 | NP_001361889.1 | 21 - 22 | 5883 | 117 | ||
NM_001375124.1 | NP_001361885.1 | 22 - 23 | 6222 | 117 | ||
NM_001375125.2 | NP_001361884.1 | 21 - 22 | 5638 | 117 | ||
NM_001383639.1 | 22 - 23 | 5766 | 117 | |||
NM_171771.6 | NP_741896.2 | 19 - 20 | 5613 | 117 | ||
GenBank mRNA | AY390550.1 | - | 12 - 13 | 1932 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |