Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313450.3 | NP_001300379.1 | 9 - 10 | 1034 | 145 | |
NM_001373541.1 | NP_001359531.1 | 9 - 10 | 1548 | 145 | ||
NM_171786.7 | NP_741913.1 | 10 - 11 | 1502 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |