Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313451.3 | NP_001300380.1 | 1 | 280 | 79 | |
NM_001373541.1 | NP_001359531.1 | 1 - 2 | 333 | 79 | ||
NM_171786.7 | NP_741913.1 | 1 - 2 | 340 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |