Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330917.4 | NP_001317908.1 | 5 - 6 | 850 | 77 | |
NM_171790.7 | NP_741917.1 | 8 - 9 | 1168 | 77 | ||
GenBank mRNA | U93195.1 | - | 8 - 9 | 1166 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |