Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383668.1 | 4 - 5 | 569 | 82 | ||
NM_171794.6 | NP_741923.1 | 3 - 4 | 430 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |