Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029268.5 | NP_001024439.1 | 6 - 7 | 643 | 70 | |
NM_171796.5 | NP_741925.2 | 2 - 3 | 486 | 70 | ||
GenBank mRNA | AJ133838.1 | - | 2 - 3 | 211 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |