Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375186.2 | NP_001362159.1 | 1 - 2 | 115 | 79 | |
NM_001375187.2 | NP_001362160.1 | 1 - 2 | 115 | 79 | ||
NM_001375188.2 | NP_001361843.1 | 2 - 3 | 248 | 79 | ||
NM_001375189.1 | NP_001361880.1 | 2 - 3 | 248 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |