Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171808.5 | NP_741941.1 | 2 - 3 | 250 | 71 | |
GenBank mRNA | U33919.1 | - | 2 - 3 | 272 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |