Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171812.7 | NP_741945.3 | 1 - 2 | 209 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |