Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wwp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306343.3 | NP_001293272.1 | 1 - 2 | 216 | 104 | |
NM_171831.4 | NP_740776.1 | 5 - 6 | 1093 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |