Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381473.2 | NP_001368649.1 | 4 - 5 | 842 | 89 | |
NM_001381474.1 | NP_001368650.1 | 4 - 5 | 776 | 89 | ||
NM_001381475.2 | NP_001368651.1 | 5 - 6 | 928 | 89 | ||
NM_001381476.1 | NP_001368652.1 | 4 - 5 | 848 | 89 | ||
NM_001381479.1 | NP_001368654.1 | 4 - 5 | 834 | 89 | ||
NM_001381480.1 | NP_001368655.1 | 4 - 5 | 839 | 89 | ||
NM_001381481.1 | NP_001368656.1 | 4 - 5 | 767 | 89 | ||
NM_171852.2 | NP_741018.1 | 5 - 6 | 898 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |