Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lon-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379712.1 | NP_001367873.1 | 4 - 5 | 566 | 105 | |
NM_065765.9 | NP_498166.1 | 4 - 5 | 564 | 105 | ||
NM_065766.7 | NP_498167.1 | 4 - 5 | 566 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |