Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027681.8 | NP_001022852.1 | 10 - 11 | 1795 | 133 | |
NM_001379977.1 | NP_001366945.1 | 11 - 12 | 1879 | 133 | ||
NM_001383020.1 | 10 - 11 | 1786 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |