Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028218.6 | NP_001023389.1 | 2 - 3 | 234 | 126 | |
NM_001028219.3 | NP_001023390.1 | 2 - 3 | 234 | 126 | ||
GenBank mRNA | AF170796.1 | - | 2 - 3 | 223 | 126 | |
AF308447.1 | - | 2 - 3 | 223 | 126 | ||
AF308448.1 | - | 2 - 3 | 223 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |