Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028998.4 | NP_001024169.1 | 7 - 8 | 1248 | 66 | |
NM_001028999.5 | NP_001024170.1 | 7 - 8 | 1599 | 66 | ||
NM_001029004.5 | NP_001024175.1 | 6 - 7 | 1022 | 66 | ||
GenBank mRNA | AF427475.1 | - | 7 - 8 | 1270 | 66 | |
AF427477.1 | - | 7 - 8 | 1453 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |