Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mlk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313223.3 | NP_001300152.1 | 11 | 1946 | 78 | |
NM_171928.6 | NP_741537.1 | 12 - 13 | 2215 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |