Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028474.7 | NP_001023645.1 | 3 - 4 | 455 | 79 | |
NM_001028475.4 | NP_001023646.1 | 3 - 4 | 461 | 79 | ||
NM_001028476.6 | NP_001023647.1 | 3 - 4 | 461 | 79 | ||
GenBank mRNA | AF072379.1 | - | 3 - 4 | 455 | 79 | |
AF072380.1 | - | 3 - 4 | 455 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |